Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234861.2 | XP_006234923.1 | 1 - 2 | 110 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |