Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108588.1 | NP_001102058.1 | 7 - 8 | 1354 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |