Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnpo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100692.1 | NP_001094162.1 | 2 - 3 | 197 | 134 | |
XM_006231839.2 | XP_006231901.1 | 3 - 4 | 788 | 134 | ||
XM_006231840.2 | XP_006231902.1 | 3 - 4 | 512 | 134 | ||
XM_008760668.1 | XP_008758890.1 | 3 - 4 | 800 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |