Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvrl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109076.1 | NP_001102546.1 | 3 - 4 | 1200 | 61 | |
XM_006250256.2 | XP_006250318.1 | 3 - 4 | 1169 | 61 | ||
GenBank mRNA | FQ228572.1 | - | 2 - 3 | 723 | 61 | |
FQ228642.1 | - | 2 - 3 | 723 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |