Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109278.2 | NP_001102748.1 | 3 - 4 | 290 | 82 | |
GenBank mRNA | FN806631.1 | - | 1 - 2 | 59 | 82 | |
FN806632.1 | - | 3 - 4 | 287 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |