Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105819.1 | NP_001099289.1 | 7 - 8 | 1133 | 77 | |
XM_006246980.2 | XP_006247042.1 | 6 - 7 | 1170 | 77 | ||
XM_006246981.1 | XP_006247043.1 | 7 - 8 | 1859 | 77 | ||
XM_006246982.2 | XP_006247044.1 | 7 - 8 | 1195 | 77 | ||
XM_008767963.1 | XP_008766185.1 | 6 - 7 | 970 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |