Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mypop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109139.1 | NP_001102609.1 | 2 - 3 | 671 | 88 | |
XM_006228452.2 | XP_006228514.1 | 2 - 3 | 1670 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus Probe spans exons |