Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182843.1 | NP_878263.1 | 2 - 3 | 475 | 65 | |
GenBank mRNA | AY336075.1 | - | 2 - 3 | 475 | 65 | |
BC083877.1 | - | 2 - 3 | 436 | 7130229 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |