Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233875.2 | XP_006233937.1 | 1 - 2 | 402 | 89 | |
XM_006233877.1 | XP_006233939.1 | 1 - 2 | 402 | 89 | ||
GenBank mRNA | U03414.1 | - | 1 - 2 | 305 | 89 | |
U03416.1 | - | 1 - 2 | 304 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |