Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767836.1 | XP_008766058.1 | 3 - 4 | 1628 | 82 | |
GenBank mRNA | EF566467.1 | - | 3 - 4 | 514 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |