Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212510.1 | NP_997675.1 | 4 - 5 | 476 | 72 | |
GenBank mRNA | BC058498.1 | - | 4 - 5 | 476 | 6919102 | 72 |
FQ209874.1 | - | 3 - 4 | 317 | 72 | ||
FQ209881.1 | - | 4 - 5 | 470 | 72 | ||
FQ209956.1 | - | 4 - 5 | 476 | 72 | ||
FQ216877.1 | - | 4 - 5 | 469 | 72 | ||
FQ221165.1 | - | 4 - 5 | 471 | 72 | ||
FQ221413.1 | - | 4 - 5 | 470 | 72 | ||
FQ221458.1 | - | 4 - 5 | 470 | 72 | ||
FQ221831.1 | - | 4 - 5 | 477 | 72 | ||
FQ222162.1 | - | 4 - 5 | 470 | 72 | ||
FQ222289.1 | - | 4 - 5 | 470 | 72 | ||
FQ222457.1 | - | 4 - 5 | 470 | 72 | ||
FQ222474.1 | - | 4 - 5 | 474 | 72 | ||
FQ222662.1 | - | 4 - 5 | 470 | 72 | ||
FQ228440.1 | - | 4 - 5 | 470 | 72 | ||
X14181.1 | - | 4 - 5 | 470 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |