Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053864.2 | NP_446316.1 | 7 - 8 | 1025 | 71 | |
GenBank mRNA | BC060518.1 | - | 7 - 8 | 1025 | 5597843 | 71 |
U11760.1 | - | 7 - 8 | 1057 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |