Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cltb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053835.1 | NP_446287.1 | 4 - 5 | 496 | 75 | |
GenBank mRNA | FQ213983.1 | - | 4 - 5 | 586 | 75 | |
M15883.1 | - | 4 - 5 | 496 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |