Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106131.1 | NP_001099601.1 | 4 - 5 | 765 | 54 | |
XM_006254942.2 | XP_006255004.1 | 4 - 5 | 1870 | 54 | ||
XM_006254943.2 | XP_006255005.1 | 4 - 5 | 1871 | 54 | ||
XM_006254944.2 | XP_006255006.1 | 4 - 5 | 1872 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Phosphatase Rattus norvegicus Probe spans exons |