Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2af2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748763.3 | XP_003748811.1 | 1 - 2 | 148 | 80 | |
XM_006228320.2 | XP_006228382.1 | 1 - 2 | 148 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |