Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019350.2 | NP_062223.1 | 5 - 6 | 859 | 71 | |
XM_006228287.2 | XP_006228349.1 | 4 - 5 | 549 | 71 | ||
GenBank mRNA | BC092198.1 | - | 5 - 6 | 859 | 71 | |
U26402.1 | - | 5 - 6 | 662 | 71 | ||
X84884.1 | - | 4 - 5 | 612 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |