Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134545.1 | NP_001128017.1 | 5 - 6 | 713 | 77 | |
XM_006228257.1 | XP_006228319.1 | 5 - 6 | 1459 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |