Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem86b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109604.1 | NP_001103074.1 | 2 - 3 | 393 | 62 | |
XM_006228289.2 | XP_006228351.1 | 1 - 2 | 661 | 62 | ||
GenBank mRNA | BC158795.1 | - | 1 - 2 | 846 | 7314455 | 62 |
FQ219460.1 | - | 2 - 3 | 307 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons |