Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnppd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199112.1 | NP_954543.1 | 4 - 5 | 574 | 105 | |
XM_006245226.2 | XP_006245288.1 | 4 - 5 | 608 | 105 | ||
GenBank mRNA | BC061750.1 | - | 4 - 5 | 574 | 5599704 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |