Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007004.1 | NP_001007005.1 | 2 - 3 | 262 | 64 | |
XM_006245227.2 | XP_006245289.1 | 2 - 3 | 661 | 64 | ||
GenBank mRNA | BC083726.1 | - | 2 - 3 | 262 | 7191229 | 64 |
FQ212913.1 | - | 286 | 64 | |||
FQ212965.1 | - | 2 - 3 | 286 | 64 | ||
FQ213109.1 | - | 2 - 3 | 286 | 64 | ||
FQ213658.1 | - | 2 - 3 | 286 | 64 | ||
FQ213871.1 | - | 2 - 3 | 286 | 64 | ||
FQ215031.1 | - | 2 - 3 | 286 | 64 | ||
FQ215251.1 | - | 2 - 3 | 286 | 64 | ||
FQ215445.1 | - | 2 - 3 | 286 | 64 | ||
FQ215549.1 | - | 2 - 3 | 286 | 64 | ||
FQ225303.1 | - | 2 - 3 | 286 | 64 | ||
FQ225881.1 | - | 2 - 3 | 286 | 64 | ||
FQ225974.1 | - | 2 - 3 | 286 | 64 | ||
FQ226026.1 | - | 2 - 3 | 283 | 64 | ||
FQ230256.1 | - | 2 - 3 | 286 | 64 | ||
FQ230321.1 | - | 2 - 3 | 286 | 64 | ||
FQ231447.1 | - | 2 - 3 | 279 | 64 | ||
FQ232744.1 | - | 2 - 3 | 286 | 64 | ||
FQ232854.1 | - | 2 - 3 | 283 | 64 | ||
FQ234185.1 | - | 2 - 3 | 286 | 64 | ||
FQ234507.1 | - | 2 - 3 | 286 | 64 | ||
FQ235001.1 | - | 2 - 3 | 286 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |