Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002730058.2 | XP_002730104.2 | 6 - 7 | 931 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |