Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108229.1 | NP_001101699.1 | 2 - 3 | 640 | 92 | |
XM_006245388.2 | XP_006245450.1 | 2 - 3 | 869 | 92 | ||
XM_006245389.2 | XP_006245451.1 | 2 - 3 | 1016 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |