Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sphkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127492.1 | NP_001120964.1 | 10 - 11 | 4809 | 90 | |
XM_006245230.2 | XP_006245292.1 | 9 - 10 | 5038 | 90 | ||
XM_006245231.2 | XP_006245293.1 | 9 - 10 | 5439 | 90 | ||
XM_006245233.2 | XP_006245295.1 | 8 - 9 | 4958 | 90 | ||
XM_008767251.1 | XP_008765473.1 | 8 - 9 | 5360 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |