Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106921.1 | NP_001100391.1 | 12 - 13 | 1475 | 65 | |
XM_008767222.1 | XP_008765444.1 | 12 - 13 | 1506 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |