Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106921.1 | NP_001100391.1 | 4 - 5 | 740 | 72 | |
XM_008767222.1 | XP_008765444.1 | 4 - 5 | 783 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Rattus norvegicus Probe spans exons |