Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021740.1 | NP_068508.1 | 3 - 4 | 359 | 105 | |
GenBank mRNA | BC061972.1 | - | 3 - 4 | 369 | 5623463 | 105 |
BC092569.1 | - | 3 - 4 | 379 | 7307282 | 105 | |
FQ210626.1 | - | 3 - 4 | 390 | 105 | ||
FQ212611.1 | - | 3 - 4 | 390 | 105 | ||
FQ214221.1 | - | 3 - 4 | 384 | 105 | ||
FQ215633.1 | - | 3 - 4 | 390 | 105 | ||
FQ219686.1 | - | 3 - 4 | 390 | 105 | ||
FQ220677.1 | - | 3 - 4 | 391 | 105 | ||
FQ221801.1 | - | 3 - 4 | 390 | 105 | ||
FQ225103.1 | - | 3 - 4 | 394 | 105 | ||
FQ226916.1 | - | 3 - 4 | 390 | 105 | ||
FQ226946.1 | - | 3 - 4 | 390 | 105 | ||
FQ227002.1 | - | 3 - 4 | 390 | 105 | ||
FQ227835.1 | - | 3 - 4 | 390 | 105 | ||
FQ229930.1 | - | 3 - 4 | 390 | 105 | ||
FQ231540.1 | - | 3 - 4 | 390 | 105 | ||
FQ231794.1 | - | 3 - 4 | 396 | 105 | ||
FQ233285.1 | - | 3 - 4 | 396 | 105 | ||
M20035.1 | - | 3 - 4 | 386 | 105 | ||
M33024.1 | - | 3 - 4 | 386 | 105 | ||
M60664.1 | - | 3 - 4 | 105 | |||
M60665.1 | - | 3 - 4 | 398 | 105 | ||
M86564.1 | - | 3 - 4 | 359 | 105 | ||
X55326.1 | - | 3 - 4 | 359 | 105 |
Gene Symbol: LOC100359583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751278.3 | XP_003751326.1 | 1 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |