Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC287004 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110636.1 | 3 - 4 | 473 | 126 | |||
NR_110637.1 | 1 - 2 | 2618 | 126 | ||||
GenBank mRNA | AY035343.1 | - | LIT1742 PINC |
3 - 4 | 470 | 126 | |
DQ080211.1 | - | LIT3459 PINC |
1 - 2 | 2618 | 126 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transition protein 1 | Tnp1 | -385.5kb | Sense | Chr. 9 - 80294837 - 80295446 on NCBI Build Rnor_6.0 | |
Downstream | tensin 1 | Tns1 | Tns | 503.9kb | Sense | Chr. 9 - 81191792 - 81401027 on NCBI Build Rnor_6.0 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |