Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012004.2 | NP_001012004.2 | 6 - 7 | 431 | 78 | |
XM_006245172.2 | XP_006245234.1 | 6 - 7 | 462 | 78 | ||
XM_006245173.1 | XP_006245235.1 | 6 - 7 | 1312 | 78 | ||
GenBank mRNA | CB691642.1 | - | 2 - 3 | 100 | 78 | |
CF114855.1 | - | 6 - 7 | 441 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |