Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100516.1 | NP_001093986.1 | 1 - 2 | 1797 | 115 | |
GenBank mRNA | CB711879.1 | - | 1 - 2 | 315 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |