Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271327.1 | NP_001258256.1 | 15 - 16 | 2644 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |