Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271327.1 | NP_001258256.1 | 7 - 8 | 1297 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |