Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289941.1 | NP_001276870.1 | 5 - 6 | 1005 | 85 | |
NM_001289942.1 | NP_001276871.1 | 6 - 7 | 1132 | 85 | ||
NM_019225.2 | NP_062098.1 | 6 - 7 | 1122 | 85 | ||
GenBank mRNA | AF265360.1 | - | 5 - 6 | 844 | 85 | |
S59158.1 | - | 6 - 7 | 1064 | 85 | ||
S75687.1 | - | 2 - 3 | 348 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |