Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csgalnact1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107309.1 | NP_001100779.1 | 3 - 4 | 953 | 117 | |
XM_006253026.2 | XP_006253088.1 | 4 - 5 | 1409 | 117 | ||
XM_006253027.2 | XP_006253089.1 | 5 - 6 | 1504 | 117 | ||
XM_006253028.2 | XP_006253090.1 | 5 - 6 | 1918 | 117 | ||
XM_006253029.2 | XP_006253091.1 | 5 - 6 | 1724 | 117 | ||
XM_006253030.2 | XP_006253092.1 | 5 - 6 | 1808 | 117 | ||
XM_006253031.2 | XP_006253093.1 | 4 - 5 | 1754 | 117 | ||
XM_008771110.1 | XP_008769332.1 | 5 - 6 | 1538 | 117 | ||
XM_008771111.1 | XP_008769333.1 | 5 - 6 | 1543 | 117 | ||
XM_008771112.1 | XP_008769334.1 | 5 - 6 | 1544 | 117 | ||
XM_008771113.1 | XP_008769335.1 | 5 - 6 | 1543 | 117 | ||
XM_008771114.1 | XP_008769336.1 | 5 - 6 | 1542 | 117 | ||
XM_008771115.1 | XP_008769337.1 | 7 - 8 | 1678 | 117 | ||
XM_008771116.1 | XP_008769338.1 | 5 - 6 | 1682 | 117 | ||
GenBank mRNA | BC168857.1 | - | 3 - 4 | 1177 | 7459880 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |