Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy5r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253014.2 | XP_006253076.1 | 1 - 2 | 155 | 91 | |
GenBank mRNA | AF044264.1 | - | 1 - 2 | 168 | 91 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |