Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033924.1 | NP_001029096.1 | 4 - 5 | 310 | 103 | |
XM_008768939.1 | XP_008767161.1 | 3 - 4 | 474 | 103 | ||
GenBank mRNA | AF381978.1 | - | 3 - 4 | 196 | 103 | |
BC078749.1 | - | 4 - 5 | 310 | 7124300 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |