Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxadrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199406.2 | NP_955438.1 | 5 - 6 | 876 | 76 | |
XM_008766539.1 | XP_008764761.1 | 5 - 6 | 1663 | 76 | ||
XM_008766540.1 | XP_008764762.1 | 4 - 5 | 983 | 76 | ||
GenBank mRNA | BC063181.1 | - | 5 - 6 | 881 | 6887452 | 76 |
Gene Symbol: LOC103690070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766579.1 | XP_008764801.1 | 1663 | 76 | ||
XM_008766580.1 | XP_008764802.1 | 1663 | 76 | |||
XM_008766581.1 | XP_008764803.1 | 983 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |