Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01403711_m1
See other TFDP2 GE Assays ›
Gene Symbol:
Tfdp2
Gene Name:
transcription factor Dp-2 (E2F dimerization partner 2)
Gene Aliases:
Tcfdp2
Chromosome Location:
Chr.8: 104040796 - 104181228 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01403711_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Tfdp2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001106847.2
NP_001100317.2
10 - 11
1188
123
XM_006243556.2
XP_006243618.1
1492
123
XM_006243558.2
XP_006243620.1
10 - 11
2591
123
XM_006243561.2
XP_006243623.1
9 - 10
2551
123
XM_006243563.1
XP_006243625.1
12 - 13
1600
123
XM_006243566.2
XP_006243628.1
1793
123
XM_006243567.2
XP_006243629.1
11 - 12
2915
123
XM_006243568.2
XP_006243630.1
12 - 13
3012
123
XM_006243569.2
XP_006243631.1
11 - 12
1726
123
XM_006243571.2
XP_006243633.1
11 - 12
1679
123
XM_006243572.2
XP_006243634.1
9 - 10
2547
123
XM_008766479.1
XP_008764701.1
11 - 12
1170
123
XM_008766480.1
XP_008764702.1
11 - 12
1166
123
XM_008766481.1
XP_008764703.1
11 - 12
1170
123
XM_008766482.1
XP_008764704.1
15 - 16
1707
123
XM_008766483.1
XP_008764705.1
11 - 12
1493
123
XM_008766484.1
XP_008764706.1
15 - 16
1561
123
XM_008766485.1
XP_008764707.1
14 - 15
1465
123
XM_008766487.1
XP_008764709.1
12 - 13
1296
123
XM_008766488.1
XP_008764710.1
9 - 10
1006
123
GenBank mRNA
BC168961.1
-
10 - 11
1188
7444676
123
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0800001331.rn.1
Set Membership:
Amplicon length greater than or equal to 101
Apoptosis
Mammalian Gene Collection
Rattus norvegicus
Probe spans exons
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cell cycle
transcription, DNA-templated
regulation of transcription, DNA-templated
heart development
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription cofactor activity
transcription factor binding
protein domain specific binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR0800009802.rn.1
TC0800001331.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0