Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ip6k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053316.2 | NP_445768.1 | 4 - 5 | 765 | 62 | |
XM_006243839.1 | XP_006243901.1 | 4 - 5 | 967 | 62 | ||
GenBank mRNA | AB049151.1 | - | 4 - 5 | 816 | 62 | |
BC078702.1 | - | 4 - 5 | 765 | 7132706 | 62 | |
FQ211545.1 | - | 4 - 5 | 913 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |