Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191885.1 | NP_001178814.1 | 6 - 7 | 1053 | 73 | |
GenBank mRNA | FQ211779.1 | - | 1013 | 73 | ||
FQ217318.1 | - | 953 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |