Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmnat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013224.1 | NP_001013242.1 | 2 - 3 | 255 | 103 | |
XM_006243608.2 | XP_006243670.1 | 2 - 3 | 585 | 103 | ||
XM_006243609.2 | XP_006243671.1 | 2 - 3 | 589 | 103 | ||
XM_006243610.2 | XP_006243672.1 | 2 - 3 | 205 | 103 | ||
GenBank mRNA | BC083725.1 | - | 2 - 3 | 255 | 7191168 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |