Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmnat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243608.2 | XP_006243670.1 | 3 - 4 | 773 | 68 | |
XM_006243609.2 | XP_006243671.1 | 3 - 4 | 777 | 68 | ||
XM_006243610.2 | XP_006243672.1 | 3 - 4 | 393 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |