Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A4gnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766588.1 | XP_008764810.1 | 2 - 3 | 628 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |