Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6gal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147205.2 | NP_671738.2 | 1 - 2 | 137 | 102 | |
GenBank mRNA | M73987.1 | - | 1 - 2 | 174 | 102 | |
M83143.1 | - | 1 - 2 | 137 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus 1700 Probe spans exons |