Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107080.2 | NP_001100550.1 | 4 - 5 | 526 | 104 | |
XM_006248609.2 | XP_006248671.1 | 4 - 5 | 564 | 104 | ||
GenBank mRNA | BC158828.1 | - | 4 - 5 | 526 | 7378165 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |