Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105864.1 | NP_001099334.1 | 3 - 4 | 406 | 90 | |
XM_006248688.2 | XP_006248750.1 | 3 - 4 | 394 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |