Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106930.1 | NP_001100400.1 | 4 - 5 | 597 | 109 | |
XM_008768871.1 | XP_008767093.1 | 4 - 5 | 628 | 109 | ||
GenBank mRNA | FQ216783.1 | - | 3 - 4 | 473 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |