Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fetub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053348.1 | NP_445800.1 | 2 - 3 | 457 | 78 | |
XM_006248549.1 | XP_006248611.1 | 3 - 4 | 411 | 78 | ||
XM_006248550.1 | XP_006248612.1 | 3 - 4 | 774 | 78 | ||
GenBank mRNA | AJ242926.1 | - | 2 - 3 | 457 | 78 | |
BC070508.1 | - | 3 - 4 | 396 | 7099154 | 78 | |
FQ209398.1 | - | 2 - 3 | 78 | |||
FQ209449.1 | - | 3 - 4 | 392 | 78 | ||
FQ209518.1 | - | 2 - 3 | 408 | 78 | ||
FQ209610.1 | - | 2 - 3 | 443 | 78 | ||
FQ209832.1 | - | 2 - 3 | 408 | 78 | ||
FQ210294.1 | - | 2 - 3 | 408 | 78 | ||
FQ210335.1 | - | 2 - 3 | 409 | 78 | ||
FQ211050.1 | - | 2 - 3 | 408 | 78 | ||
FQ218476.1 | - | 2 - 3 | 439 | 78 | ||
FQ218557.1 | - | 3 - 4 | 387 | 78 | ||
FQ218709.1 | - | 2 - 3 | 408 | 78 | ||
FQ218884.1 | - | 3 - 4 | 379 | 78 | ||
FQ218893.1 | - | 2 - 3 | 408 | 78 | ||
FQ218905.1 | - | 2 - 3 | 408 | 78 | ||
FQ218930.1 | - | 2 - 3 | 441 | 78 | ||
FQ218949.1 | - | 2 - 3 | 258 | 78 | ||
FQ219048.1 | - | 2 - 3 | 439 | 78 | ||
FQ219152.1 | - | 2 - 3 | 445 | 78 | ||
FQ219170.1 | - | 2 - 3 | 408 | 78 | ||
FQ219190.1 | - | 2 - 3 | 443 | 78 | ||
FQ219266.1 | - | 2 - 3 | 408 | 78 | ||
FQ219352.1 | - | 2 - 3 | 408 | 78 | ||
FQ219454.1 | - | 2 - 3 | 408 | 78 | ||
FQ219459.1 | - | 2 - 3 | 406 | 78 | ||
FQ219500.1 | - | 2 - 3 | 411 | 78 | ||
FQ219513.1 | - | 2 - 3 | 480 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |