Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fetub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053348.1 | NP_445800.1 | 4 - 5 | 708 | 92 | |
XM_006248549.1 | XP_006248611.1 | 5 - 6 | 662 | 92 | ||
XM_006248550.1 | XP_006248612.1 | 5 - 6 | 1025 | 92 | ||
GenBank mRNA | AJ242926.1 | - | 4 - 5 | 708 | 92 | |
BC070508.1 | - | 5 - 6 | 647 | 7099154 | 92 | |
FQ209610.1 | - | 4 - 5 | 93 | |||
FQ209832.1 | - | 4 - 5 | 660 | 92 | ||
FQ210294.1 | - | 4 - 5 | 659 | 92 | ||
FQ211050.1 | - | 4 - 5 | 659 | 92 | ||
FQ211248.1 | - | 4 - 5 | 694 | 92 | ||
FQ218476.1 | - | 4 - 5 | 690 | 92 | ||
FQ218709.1 | - | 4 - 5 | 659 | 93 | ||
FQ218884.1 | - | 5 - 6 | 630 | 92 | ||
FQ218930.1 | - | 4 - 5 | 92 | |||
FQ218949.1 | - | 4 - 5 | 509 | 92 | ||
FQ219048.1 | - | 4 - 5 | 690 | 92 | ||
FQ219152.1 | - | 4 - 5 | 696 | 92 | ||
FQ219170.1 | - | 4 - 5 | 659 | 92 | ||
FQ219190.1 | - | 4 - 5 | 694 | 92 | ||
FQ219266.1 | - | 4 - 5 | 659 | 92 | ||
FQ219352.1 | - | 4 - 5 | 659 | 92 | ||
FQ219426.1 | - | 2 - 3 | 261 | 92 | ||
FQ219454.1 | - | 4 - 5 | 659 | 92 | ||
FQ219459.1 | - | 4 - 5 | 92 | |||
FQ219500.1 | - | 4 - 5 | 662 | 92 | ||
FQ219513.1 | - | 4 - 5 | 731 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |