Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105863.2 | NP_001099333.2 | 4 - 5 | 411 | 75 | |
XM_008768864.1 | XP_008767086.1 | 3 - 4 | 394 | 75 | ||
GenBank mRNA | AW916308.1 | - | 4 - 5 | 411 | 75 | |
BC083814.1 | - | 1 - 2 | 2049 | 75 | ||
BC168998.1 | - | 3 - 4 | 333 | 7387094 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |