Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102418.1 | NP_001095888.1 | 6 - 7 | 868 | 72 | |
NM_012741.1 | NP_036873.1 | 6 - 7 | 769 | 72 | ||
GenBank mRNA | BC088155.1 | - | 6 - 7 | 868 | 7369450 | 72 |
L29428.1 | - | 6 - 7 | 769 | 72 | ||
M11884.1 | - | 4 - 5 | 423 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |